2-(2-Pyridyl)benzoxazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyridin-2-yl)benzo[d]oxazole

A467363-1G A467363-250MG A467363-25G A467363-5G
77102-694EA 47.75 USD
77102-694 77102-696 77102-698 77102-700
2-(2-Pyridyl)benzoxazole 97%
2-(2-Pyridyl)benzoxazole

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O
MW: 196.21 g/mol
MDL Number: MFCD00569582
CAS Number: 32959-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR