5-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
A467334-1G
A467334-250MG
A467334-5G
77102-688EA
95
USD
77102-688
77102-690
77102-692
5-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester 98%
5-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₇H₂₅BFNO₄ |
MDL Number:
MFCD22419260 CAS Number: 1449135-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...