(S)-3-Amino-2-(((benzyloxy)carbonyl)amino)propanoic acid 97%

Supplier: Ambeed
Warning

A466981-1G A466981-5G A466981-100G A466981-10G A466981-25G
77102-684EA 5 USD
77102-684 77102-686 77613-438 77613-440 77686-243
(S)-3-Amino-2-(((benzyloxy)carbonyl)amino)propanoic acid 97%
(S)-3-Amino-2-(((benzyloxy)carbonyl)amino)propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₄
MDL Number: MFCD00237345
CAS Number: 35761-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR