3-Bromo-4,5-diethoxy-benzylamine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A466161-1G
A466161-250MG
77102-678EA
926.17
USD
77102-678
77102-680
3-Bromo-4,5-diethoxy-benzylamine hydrochloride 95%
3-Bromo-4,5-diethoxy-benzylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₇BrClNO₂ MW: 310.61 g/mol |
MDL Number:
MFCD06634747 CAS Number: 1052510-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...