(1,2,3-Thiadiazol-4-yl)methanol 95%

Supplier: Ambeed
Danger

A465796-5G A465796-1G
77102-676EA 571 USD
77102-676 77686-241
(1,2,3-Thiadiazol-4-yl)methanol 95%
(1,2,3-Thiadiazol-4-yl)methanol

Precaución: for Research Use Only

Formula: C₃H₄N₂OS
MDL Number: MFCD06202704
CAS Number: 50868-99-0
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR