6-Methoxy-1H-indole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A463369-100MG A463369-1G A463369-250MG
77102-656EA 152.77 USD
77102-656 77102-658 77102-660
6-Methoxy-1H-indole-2-carbaldehyde 95%
6-Methoxy-1H-indole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.18 g/mol
MDL Number: MFCD01545314
CAS Number: 30464-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR