Fmoc-D-Cys(Mob)-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((4-methoxybenzyl)thio)propanoic acid

A462730-1G A462730-100MG A462730-250MG A462730-5G
77102-652EA 10 USD
77102-652 77901-559 77901-954 77900-511
Fmoc-D-Cys(Mob)-OH 95%
Fmoc-D-Cys(Mob)-OH

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₅S
MDL Number: MFCD00237015
CAS Number: 200354-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR