4-tert-Butyl-1-(bromomethyl)benzene 98%

Supplier: Ambeed
Danger

A462583-10G A462583-25G A462583-5G
77102-646EA 12 USD
77102-646 77102-648 77102-650
4-tert-Butyl-1-(bromomethyl)benzene 98%
4-tert-Butyl-1-(bromomethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅Br
MDL Number: MFCD00000180
CAS Number: 18880-00-7
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR