3-Amino-5-fluoropyridine-2-carbonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 3-Amino-2-cyano-5-fluoropyridine, 3-Amino-5-fluoropicolinonitrile

A461326-100MG A461326-1G A461326-250MG
77102-640EA 109.28 USD
77102-640 77102-642 77102-644
3-Amino-5-fluoropyridine-2-carbonitrile 95%
3-Amino-5-fluoropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₄FN₃
MW: 137.114 g/mol
MDL Number: MFCD20259119
CAS Number: 1374115-60-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR