2-Methoxy-7-methylnaphthalene 95+%

Supplier: Ambeed
Warning

A461246-100MG A461246-250MG
77102-636EA 39.26 USD
77102-636 77102-638
2-Methoxy-7-methylnaphthalene 95+%
2-Methoxy-7-methylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
MW: 172.223 g/mol
MDL Number: MFCD18412839
CAS Number: 1131-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR