(S)-Methyl 2-amino-3-((tert-butoxycarbonyl)amino)-3-methylbutanoate 95%

Supplier: Ambeed
Warning

A461085-1G A461085-250MG A461085-100MG
77102-632EA 371.32 USD
77102-632 77102-634 77686-235
(S)-Methyl 2-amino-3-((tert-butoxycarbonyl)amino)-3-methylbutanoate 95%
(S)-Methyl 2-amino-3-((tert-butoxycarbonyl)amino)-3-methylbutanoate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₄
MW: 246.30 g/mol
MDL Number: MFCD16293742
CAS Number: 1093192-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR