2,5-Difluorobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A460408-100G A460408-1G A460408-25G A460408-5G
77102-624EA 208 USD
77102-624 77102-626 77102-628 77102-630
2,5-Difluorobenzenesulfonamide 98%
2,5-Difluorobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₅F₂NO₂S
MDL Number: MFCD00798115
CAS Number: 120022-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR