1-Bromo-2,4-difluorobenzene 97%

Supplier: Ambeed
Danger

Synonyms: 2,4-Difluorobromobenzene

A460226-25G A460226-5G A460226-100G
77102-618EA 8.49 USD
77102-618 77102-620 77594-446
1-Bromo-2,4-difluorobenzene 97%
1-Bromo-2,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrF₂
MW: 192.99 g/mol
MDL Number: MFCD00000330
CAS Number: 348-57-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR