2-(1-Pyrazolyl)benzyl alcohol 95%

Supplier: Ambeed
Warning

A459206-1G A459206-250MG A459206-5G
77102-588EA 70 USD
77102-588 77102-590 77102-592
2-(1-Pyrazolyl)benzyl alcohol 95%
2-(1-Pyrazolyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD03086188
CAS Number: 741717-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR