Ethyl 2-fluoro-4-methylbenzoate 98+%

Supplier: Ambeed
Warning

A457376-1G A457376-250MG
77102-562EA 13 USD
77102-562 77102-564
Ethyl 2-fluoro-4-methylbenzoate 98+%
Ethyl 2-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MDL Number: MFCD06205105
CAS Number: 500579-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR