5,6-Dichloro-3-iodo-1H-indazole 95%

Supplier: Ambeed
Warning

A456750-1G A456750-250MG
77102-556EA 183.54 USD
77102-556 77102-558
5,6-Dichloro-3-iodo-1H-indazole 95%
5,6-Dichloro-3-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₃Cl₂IN₂
MW: 312.92 g/mol
MDL Number: MFCD26793730
CAS Number: 1426421-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR