PFI-3 99+%

Supplier: Ambeed
Warning

A455988-10MG A455988-1MG A455988-50MG A455988-5MG
77102-542EA 85 USD
77102-542 77102-544 77102-546 77102-548
PFI-3 99+%
PFI-3

Precaución: for Research Use Only

Formula: C₁₉H₁₉N₃O₂
MDL Number: MFCD28100807
CAS Number: 1819363-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR