6-Aminobenzimidazole 97%

Supplier: Ambeed
Warning

A453802-1G A453802-25G A453802-5G A453802-10G
77102-536EA 14.86 USD
77102-536 77102-538 77102-540 77594-426
6-Aminobenzimidazole 97%
6-Aminobenzimidazole

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD00831692
CAS Number: 934-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR