Ethyl malonyl chloride 90%, Technical Grade

Supplier: Ambeed
Danger

Synonyms: Ethyl Malonyl Chloride

A451975-10G A451975-25G A451975-5G A451975-100G
77102-530EA 19.1 USD
77102-530 77102-532 77102-534 77594-410
Ethyl malonyl chloride 90%, Technical Grade
Ethyl malonyl chloride

Precaución: for Research Use Only

Formula: C₅H₇ClO₃
MW: 150.56 g/mol
MDL Number: MFCD00000736
CAS Number: 36239-09-5
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR