2,6-Dichloro-3-(trifluoromethyl)benzyl bromide 95%

Supplier: Ambeed
Danger

A450701-1G A450701-250MG
77102-522EA 58 USD
77102-522 77102-524
2,6-Dichloro-3-(trifluoromethyl)benzyl bromide 95%
2,6-Dichloro-3-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₄BrCl₂F₃
MDL Number: MFCD11519349
CAS Number: 1092461-07-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR