(E)-Methyl 3-(3-methoxyphenyl)acrylate 95%

Supplier: Ambeed
Warning

A450597-1G A450597-5G
77102-518EA 28.65 USD
77102-518 77102-520
(E)-Methyl 3-(3-methoxyphenyl)acrylate 95%
(E)-Methyl 3-(3-methoxyphenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
MDL Number: MFCD22381372
CAS Number: 38693-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR