2-Amino-5-bromo-N-methylbenzamide 95%

Supplier: Ambeed
Warning

A449427-100MG A449427-1G A449427-5G
77102-508EA 13 USD
77102-508 77102-510 77572-410
2-Amino-5-bromo-N-methylbenzamide 95%
2-Amino-5-bromo-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O
MDL Number: MFCD18089344
CAS Number: 1257996-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR