5-Chloro-3-methylpyrazine-2-carbonitrile 95%

Supplier: Ambeed
Warning

A448763-250MG A448763-100MG
77102-506EA 260.99 USD
77102-506 77686-212
5-Chloro-3-methylpyrazine-2-carbonitrile 95%
5-Chloro-3-methylpyrazine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.56 g/mol
MDL Number: MFCD18250189
CAS Number: 1260665-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR