2-Chloro-4-nitrophenol 98%

Supplier: Ambeed
Danger

A448613-1000G A448613-100G A448613-10G A448613-25G A448613-500G A448613-5G
77102-496EA 205 USD
77102-496 77102-498 77102-500 77102-502 77102-504 77900-805
2-Chloro-4-nitrophenol 98%
2-Chloro-4-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MDL Number: MFCD00043910
CAS Number: 619-08-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR