2-(Trifluoromethoxy)benzoyl chloride 98%

Supplier: Ambeed
Danger

A447516-1G A447516-25G A447516-5G
77102-482EA 12 USD
77102-482 77901-143 77901-870
2-(Trifluoromethoxy)benzoyl chloride 98%
2-(Trifluoromethoxy)benzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MDL Number: MFCD00052330
CAS Number: 162046-61-9
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR