9-(p-Tolyl)carbazole 98%

Supplier: Ambeed
Warning

Synonyms: 9-(p-Tolyl)-9H-carbazole

A447114-100MG A447114-1G A447114-250MG A447114-5G
77102-474EA 18.04 USD
77102-474 77102-476 77102-478 77102-480
9-(p-Tolyl)carbazole 98%
9-(p-Tolyl)carbazole

Precaución: for Research Use Only

Formula: C₁₉H₁₅N
MW: 257.34 g/mol
MDL Number: MFCD14708175
CAS Number: 19264-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR