(2S,3S)-(+)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene 98%

Supplier: Ambeed
Warning

Synonyms: (S,S)-NORPHOS, (2S,3S)-(+)-Norphos, (2S,3S)-(+)-2,3-Bis(diphenylphosphino)-bicyclo[2.2.1]hept-5-ene, (1R,4S,5S,6S)-5,6-Bis(diphenylphosphaneyl)bicyclo[2.2.1]hept-2-ene

A446594-100MG A446594-1G A446594-250MG
77102-468EA 150.65 USD
77102-468 77102-470 77102-472
(2S,3S)-(+)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene 98%
(2S,3S)-(+)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene

Precaución: for Research Use Only

Formula: C₃₁H₂₈P₂
MW: 462.51 g/mol
MDL Number: MFCD00085364
CAS Number: 71042-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR