tert-Butyl (2-(aminooxy)ethyl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: tert-butyl N-[2-(aminooxy)ethyl]carbamate

A445157-250MG A445157-1G A445157-5G
77102-456EA 18 USD
77102-456 77686-205 77686-206
tert-Butyl (2-(aminooxy)ethyl)carbamate 98%
tert-Butyl (2-(aminooxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₇H₁₆N₂O₃
MDL Number: MFCD11934722
CAS Number: 75051-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR