tert-Butyl (2-methylbut-3-yn-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A443860-100MG A443860-250MG A443860-1G
77102-452EA 30.77 USD
77102-452 77594-390 77613-364
tert-Butyl (2-methylbut-3-yn-2-yl)carbamate 95%
tert-Butyl (2-methylbut-3-yn-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₂
MW: 183.25 g/mol
MDL Number: MFCD16036595
CAS Number: 113486-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR