α'-Bromo-α,α,α-trifluoro-p-xylene 98%

Supplier: Ambeed
Danger

A439908-10G A439908-1G A439908-25G A439908-5G
77102-420EA 8 USD
77102-420 77102-422 77102-424 77102-426
α'-Bromo-α,α,α-trifluoro-p-xylene 98%
α'-Bromo-α,α,α-trifluoro-p-xylene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MDL Number: MFCD00000403
CAS Number: 402-49-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR