4,4,5,5-Tetramethyl-2-(3-(methylsulfinyl)phenyl)-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Danger

A439698-1G A439698-250MG A439698-5G
77102-414EA 56 USD
77102-414 77102-416 77102-418
4,4,5,5-Tetramethyl-2-(3-(methylsulfinyl)phenyl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(3-(methylsulfinyl)phenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₃S
MDL Number: MFCD18733605
CAS Number: 1416367-04-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR