Naphthalene-2,6-dicarbonitrile 95%

Supplier: Ambeed
Warning

A439446-100MG A439446-1G A439446-250MG A439446-5G
77102-400EA 105.03 USD
77102-400 77102-402 77102-404 77102-406
Naphthalene-2,6-dicarbonitrile 95%
Naphthalene-2,6-dicarbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₆N₂
MW: 178.18 g/mol
MDL Number: MFCD14583105
CAS Number: 31656-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR