Potassium (3-chloro-5-methoxyphenyl)trifluoroborate 96%

Supplier: Ambeed
Warning

A439114-1G A439114-250MG A439114-5G
77102-390EA 162 USD
77102-390 77102-392 77102-394
Potassium (3-chloro-5-methoxyphenyl)trifluoroborate 96%
Potassium (3-chloro-5-methoxyphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₆BClF₃KO
MW: 248.48 g/mol
MDL Number: MFCD09993766
CAS Number: 929626-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR