2,5-Dioxopyrrolidin-1-yl 4-iodobenzoate 98%

Supplier: Ambeed
Warning

A438788-1G A438788-250MG A438788-5G
77102-384EA 66.84 USD
77102-384 77102-386 77102-388
2,5-Dioxopyrrolidin-1-yl 4-iodobenzoate 98%
2,5-Dioxopyrrolidin-1-yl 4-iodobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₈INO₄
MW: 345.09 g/mol
MDL Number: MFCD24387072
CAS Number: 39028-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR