α-Bromo-3,4-dichlorotoluene 98%

Supplier: Ambeed
Danger

A437018-25G A437018-5G
77102-380EA 39 USD
77102-380 77102-382
α-Bromo-3,4-dichlorotoluene 98%
α-Bromo-3,4-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MDL Number: MFCD00041363
CAS Number: 18880-04-1
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR