3-Chloro-4-(trifluoromethoxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

A436282-1G A436282-250MG A436282-100MG A436282-5G
77102-376EA 150 USD
77102-376 77102-378 77686-187 77901-739
3-Chloro-4-(trifluoromethoxy)phenylboronic acid 98%
3-Chloro-4-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₃
MDL Number: MFCD10697423
CAS Number: 870822-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR