D-4-TBU-Phe-OH 95%

Supplier: Ambeed
Danger

Synonyms: H-D-Phe(4-tBu)-OH, (R)-2-Amino-3-(4-(tert-butyl)phenyl)propanoic acid, H-p-tBu-D-Phe-OH

A435699-250MG A435699-25G A435699-5G A435699-1G
77102-370EA 36 USD
77102-370 77102-372 77102-374 77686-186
D-4-TBU-Phe-OH 95%
D-4-TBU-Phe-OH

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD02259529
CAS Number: 274262-82-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR