2-Phenylnaphtho[2,3-d]oxazole-4,9-dione 98%

Supplier: Ambeed
Warning

Synonyms: SJB2-043

A434626-100MG A434626-10MG A434626-1MG A434626-50MG A434626-5MG
77102-348EA 240.83 USD
77102-348 77102-350 77102-352 77102-354 77102-356
2-Phenylnaphtho[2,3-d]oxazole-4,9-dione 98%
2-Phenylnaphtho[2,3-d]oxazole-4,9-dione

Precaución: for Research Use Only

Formula: C₁₇H₉NO₃
MW: 275.25 g/mol
MDL Number: MFCD26960957
CAS Number: 63388-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR