1-Chloro-2,3,4-trifluorobenzene 95%

Supplier: Ambeed
Warning

A434404-100G A434404-10G A434404-1G A434404-25G A434404-5G
77102-332EA 176.11 USD
77102-332 77102-334 77102-336 77102-338 77102-340
1-Chloro-2,3,4-trifluorobenzene 95%
1-Chloro-2,3,4-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₃
MW: 166.5300 g/mol
MDL Number: MFCD00042222
CAS Number: 36556-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR