2-Bromo-5-(difluoromethoxy)benzaldehyde 97%

Supplier: Ambeed
Warning

A433753-100MG A433753-1G A433753-250MG A433753-5G
77102-320EA 41.38 USD
77102-320 77102-322 77102-324 77102-326
2-Bromo-5-(difluoromethoxy)benzaldehyde 97%
2-Bromo-5-(difluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
MW: 251.02 g/mol
MDL Number: MFCD24131095
CAS Number: 1504952-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR