2-Methylpropane-1-sulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: Isobutanesulphonyl chloride, 1-Propanesulphonylchloride, 2-methyl-

A433732-10G A433732-1G A433732-5G A433732-25G
77102-314EA 57 USD
77102-314 77102-316 77102-318 77551-300
2-Methylpropane-1-sulfonyl chloride 97%
2-Methylpropane-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₄H₉ClO₂S
MDL Number: MFCD07369800
CAS Number: 35432-36-1
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR