5-Bromo-3-methylpyridine-2-carboxaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-3-methylpicolinaldehyde, 5-Bromo-2-formyl-3-methylpyridine, 5-Bromo-3-methyl-2-pyridinecarbaldehyde
A431910-10G
A431910-1G
A431910-250MG
A431910-5G
77102-278EA
951.63
USD
77102-278
77102-280
77102-282
77102-284
5-Bromo-3-methylpyridine-2-carboxaldehyde 97%
5-Bromo-3-methylpyridine-2-carboxaldehyde
Precaución: for Research Use Only
Formula:
C₇H₆BrNO MW: 200.033 g/mol |
MDL Number:
MFCD07776420 CAS Number: 376587-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...