2,3-Dihydro-1H-cyclopenta[b]quinolin-9-amine 98%

Supplier: Ambeed
Warning

A431459-1G A431459-250MG
77102-274EA 166.57 USD
77102-274 77102-276
2,3-Dihydro-1H-cyclopenta[b]quinolin-9-amine 98%
2,3-Dihydro-1H-cyclopenta[b]quinolin-9-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
MW: 184.23 g/mol
MDL Number: MFCD00618440
CAS Number: 18528-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR