3-(Methylamino)isonicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Methylamino)pyridine-4-carboxylic acid, 3-(Methylamino)isonicotinic acid

A431226-10G A431226-1G A431226-5G
77102-268EA 360 USD
77102-268 77102-270 77102-272
3-(Methylamino)isonicotinic acid 97%
3-(Methylamino)isonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.151 g/mol
MDL Number: MFCD24160213
CAS Number: 1461602-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR