Pentaerythritol triacrylate stabilized, Technical Grade

Supplier: Ambeed
Danger

Synonyms: Pentaerythrityl Triacrylate, 2-((Acryloyloxy)methyl)-2-(hydroxymethyl)propane-1,3-diyl diacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 200 - 600 ppm

A431091-100G A431091-25G A431091-500G A431091-1000G
77102-260EA 9.55 USD
77102-260 77102-262 77102-264 77572-336
Pentaerythritol triacrylate stabilized, Technical Grade
Pentaerythritol triacrylate

Precaución: for Research Use Only

Formula: C₁₄H₁₈O₇
MW: 298.28 g/mol
MDL Number: MFCD00009607
CAS Number: 3524-68-3
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR