5-Bromo-7-methoxy-1H-indazole 96%

Supplier: Ambeed
Warning

A430245-100MG A430245-1G A430245-250MG A430245-5G
77102-248EA 102.91 USD
77102-248 77102-250 77102-252 77686-173
5-Bromo-7-methoxy-1H-indazole 96%
5-Bromo-7-methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.05 g/mol
MDL Number: MFCD11878570
CAS Number: 1374652-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR