2-Bromo-9H-thioxanthen-9-one 95%

Supplier: Ambeed
Warning

A429076-100MG A429076-1G A429076-250MG A429076-5G
77102-234EA 20.16 USD
77102-234 77102-236 77102-238 77102-240
2-Bromo-9H-thioxanthen-9-one 95%
2-Bromo-9H-thioxanthen-9-one

Precaución: for Research Use Only

Formula: C₁₃H₇BrOS
MW: 291.16 g/mol
MDL Number: MFCD00093494
CAS Number: 20077-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR