1-(4-(Trifluoromethoxy)phenyl)propan-2-one 95%

Supplier: Ambeed
Warning

A427494-1G A427494-250MG A427494-5G
77102-208EA 39 USD
77102-208 77102-210 77102-212
1-(4-(Trifluoromethoxy)phenyl)propan-2-one 95%
1-(4-(Trifluoromethoxy)phenyl)propan-2-one

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MDL Number: MFCD03410496
CAS Number: 1249274-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR