Dimethyl-2-nitroterephthalate 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 2-nitroterephthalate

A427078-100G A427078-10G A427078-25G
77102-196EA 28 USD
77102-196 77102-198 77102-200 77102-202
Dimethyl-2-nitroterephthalate 98%
Dimethyl-2-nitroterephthalate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₆
MW: 239.18 g/mol
MDL Number: MFCD00008422
CAS Number: 5292-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR