ω-Bromo-3-chloroacetophenone 97%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-3'-chloroacetophenone, Bromomethyl m-chlorophenyl ketone, 2-Bromo-1-(3-chlorophenyl)ethanone, 3-Chlorophenacyl bromide, α-Bromo-3-chloroacetophenone

A425791-10G A425791-1G A425791-250MG A425791-25G A425791-5G
77102-184EA 23 USD
77102-184 77102-186 77102-188 77102-190 77102-192
ω-Bromo-3-chloroacetophenone 97%
ω-Bromo-3-chloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MDL Number: MFCD00792669
CAS Number: 41011-01-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR